Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001267576.1 | NP_001254505.1 | 6 | 1151 | |
NM_153210.4 | NP_694942.3 | 6 | 1151 | ||
XM_011523639.2 | XP_011521941.1 | 5 | 1039 | ||
XM_011523640.1 | XP_011521942.1 | 5 | 871 | ||
XM_011523641.1 | XP_011521943.1 | 6 | 596 | ||
XM_011523642.2 | XP_011521944.1 | 6 | 1196 | ||
XM_017024159.1 | XP_016879648.1 | 5 | 1092 | ||
XM_017024160.1 | XP_016879649.1 | 4 | 1078 | ||
XM_017024161.1 | XP_016879650.1 | 3 | 1164 | ||
XM_017024162.1 | XP_016879651.1 | 6 | 1196 | ||
GenBank mRNA | AJ583817.2 | - | 6 | 1058 | |
AK055188.1 | - | 6 | 632 | ||
BC136368.1 | - | 6 | 1151 | 9020355 | |
BC144041.1 | - | 6 | 1151 | 9052561 | |
BC171759.1 | - | 6 | 1151 | 9054412 | |
BX648526.1 | - | 4 | 414 |