Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190259.1 | NP_001177188.1 | 6 | 1462 | |
NM_001190260.1 | NP_001177189.1 | 5 | 1411 | ||
NM_152785.4 | NP_689998.1 | 6 | 1456 | ||
XM_005247272.2 | XP_005247329.1 | 6 | 1880 | ||
XM_011512627.2 | XP_011510929.1 | 6 | 1334 | ||
XM_011512628.2 | XP_011510930.1 | 6 | 1391 | ||
XM_011512629.1 | XP_011510931.1 | 6 | 1883 | ||
XM_011512630.1 | XP_011510932.1 | 6 | 1889 | ||
GenBank mRNA | AF521911.1 | - | 6 | 1443 | |
AY212246.1 | - | 6 | 1376 | ||
BC030506.1 | - | 6 | 1324 | 4385178 |