Gene Transcripts

Gene Symbol
CNTD1
Entrez Gene ID
124817
Gene Name
cyclin N-terminal domain containing 1
Gene Aliases
CNTD
Chromosome Location
Chr. 17: 42798667 - 42811587 on Build GRCh38
UniGene ID
Hs.12272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173478.2 NP_775749.2 2 465
XM_005257043.4 XP_005257100.1 2 228
XM_005257044.4 XP_005257101.1 2 246
XM_006721690.2 XP_006721753.1 2 465
XM_011524310.2 XP_011522612.1 2 412
GenBank mRNA AK097456.1 - 2 381
AK302094.1 - 2 198
BC026187.2 - 2 509 5267540

View Full Product Details