Gene Transcripts

Gene Symbol
UMODL1
Entrez Gene ID
89766
Gene Name
uromodulin like 1
Gene Aliases
Chromosome Location
Chr. 21: 42062959 - 42142996 on Build GRCh38
UniGene ID
Hs.242520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004416.2 NP_001004416.2 19 3442
NM_001199527.1 NP_001186456.1 18 4006
NM_001199528.2 NP_001186457.2 19 3622
NM_173568.3 NP_775839.3 18 3826
XM_011529797.1 XP_011528099.1 16 3434
XM_017028506.1 XP_016883995.1 18 3821
XM_017028507.1 XP_016883996.1 18 6750
XM_017028508.1 XP_016883997.1 8 1605
GenBank mRNA AB051810.1 - 18 4006
AB051811.1 - 19 3622
AK093654.1 - 4 684
AK124223.1 - 5 1186
AY771619.1 - 18 3826
AY771620.1 - 19 3442

View Full Product Details