Gene Transcripts

Gene Symbol
CRX
Entrez Gene ID
1406
Gene Name
cone-rod homeobox
Gene Aliases
CORD2, CRD, LCA7, OTX3
Chromosome Location
Chr. 19: 47821842 - 47843330 on Build GRCh38
UniGene ID
Hs.617342
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000554.4 NP_000545.1 2 277
GenBank mRNA AL711350.1 - 2 277
BC016664.1 - 2 212 3869862
BC053672.1 - 2 163 6144191
BT007364.1 - 1 73
DQ340179.1 - 1 73
DQ426868.1 - 2 217

View Full Product Details