Gene Transcripts

Gene Symbol
SLC29A4
Entrez Gene ID
222962
Gene Name
solute carrier family 29 member 4
Gene Aliases
ENT4, PMAT
Chromosome Location
Chr. 7: 5274311 - 5304073 on Build GRCh38
UniGene ID
Hs.4302
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040661.1 NP_001035751.1 4 498
NM_001300847.1 NP_001287776.1 4 495
NM_153247.2 NP_694979.2 4 495
XM_005249658.4 XP_005249715.1 4 461
XM_006715667.3 XP_006715730.1 4 366
XM_006715668.2 XP_006715731.1 4 481
XM_011515200.2 XP_011513502.1 4 526
XM_011515201.2 XP_011513503.1 4 515
GenBank mRNA AK075422.1 - 2 72
AK092242.1 - 4 481
AK296278.1 - 4 466
AL515106.3 - 4 496
AY485959.1 - 4 495
BC025325.1 - 4 396 3354432
BC047592.1 - 4 459 4816109

Gene Symbol
LOC100287294
Entrez Gene ID
100287294
Gene Name
equilibrative nucleoside transporter 4-like
Gene Aliases
Chromosome Location
Chr. 7: 56891391 - 56945473 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017012902.1 XP_016868391.1 4 406

View Full Product Details