Gene Transcripts

Gene Symbol
GOLGA3
Entrez Gene ID
2802
Gene Name
golgin A3
Gene Aliases
GCP170, MEA-2
Chromosome Location
Chr. 12: 132768909 - 132829131 on Build GRCh38
UniGene ID
Hs.507333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005895.3 NP_005886.2 24 5218
XM_005266164.4 XP_005266221.1 24 4825
XM_005266167.4 XP_005266224.1 21 4250
XM_006719736.3 XP_006719799.1 24 4945
XM_006719737.3 XP_006719800.1 24 4955
XM_011534793.2 XP_011533095.1 23 4672
XM_011534794.2 XP_011533096.1 16 2823
GenBank mRNA D63997.1 - 24 4923

View Full Product Details