Gene Transcripts

Gene Symbol
SYT2
Entrez Gene ID
127833
Gene Name
synaptotagmin 2
Gene Aliases
CMS7, MYSPC, SytII
Chromosome Location
Chr. 1: 202590596 - 202710444 on Build GRCh38
UniGene ID
Hs.25422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK090672.1 - 2 299
AK094430.1 - 2 315
BC100817.1 - 2 170 40001000
DA098279.1 - 2 308

View Full Product Details