Gene Transcripts

Gene Symbol
SULT1C2
Entrez Gene ID
6819
Gene Name
sulfotransferase family 1C member 2
Gene Aliases
ST1C1, ST1C2, SULT1C1, humSULTC2
Chromosome Location
Chr. 2: 108288639 - 108309915 on Build GRCh38
UniGene ID
Hs.436123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001056.3 NP_001047.1 2 579
NM_176825.2 NP_789795.1 2 579
XM_005264013.3 XP_005264070.1 2 394
GenBank mRNA AB008164.1 - 2 249
AF026303.1 - 2 211
AF186251.1 - 2 444
AF186252.1 - 2 579
AF186253.1 - 2 295
AF186254.1 - 2 444
AF186255.1 - 2 579
AF186256.1 - 2 295
AK297086.1 - 2 343
AK298306.1 - 2 312
AK313193.1 - 2 343
BC005353.1 - 2 196 4051264
BT006951.1 - 1 126
U66036.1 - 2 302

View Full Product Details