Gene Transcripts

Gene Symbol
PDLIM2
Entrez Gene ID
64236
Gene Name
PDZ and LIM domain 2
Gene Aliases
MYSTIQUE, SLIM
Chromosome Location
Chr. 8: 22578741 - 22598025 on Build GRCh38
UniGene ID
Hs.632034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021630.5 NP_067643.3 5 1123
NM_176871.3 NP_789847.1 4 465
NM_198042.3 NP_932159.1 4 465
GenBank mRNA AF289563.1 - 5 722
AK024498.1 - 4 408
AK027800.1 - 5 734
AK092968.1 - 5 1123
AY007729.1 - 4 489
AY070438.1 - 4 489
AY217349.1 - 4 450
BC021556.1 - 5 604 3959372
BC071774.1 - 5 734 4593207
DN997519.1 - 5 409

View Full Product Details