Gene Transcripts

Gene Symbol
TAS2R50
Entrez Gene ID
259296
Gene Name
taste 2 receptor member 50
Gene Aliases
T2R50, T2R51, TAS2R51
Chromosome Location
Chr. 12: 10985913 - 10986912 on Build GRCh38
UniGene ID
Hs.688194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176890.2 NP_795371.2 1 855
GenBank mRNA BC112100.1 - 1 855

View Full Product Details