Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300860.1 | NP_001287789.1 | 2 | 284 | |
NM_014829.3 | NP_055644.2 | 2 | 284 | ||
NR_125341.1 | 2 | 284 | |||
XM_011543758.1 | XP_011542060.1 | 2 | 247 | ||
GenBank mRNA | AB018344.1 | - | 2 | 223 | |
AF106680.1 | - | 2 | 227 | ||
AK126011.1 | - | 2 | 239 | ||
AK291788.1 | - | 2 | 237 | ||
BC012304.2 | - | 2 | 214 | 3873814 | |
BC105598.1 | - | 2 | 241 | ||
BF244218.1 | - | 2 | 285 |