Gene Transcripts
- Gene Symbol
- ACYP2
- Entrez Gene ID
- 98
- Gene Name
- acylphosphatase 2
- Gene Aliases
- ACYM, ACYP
- Chromosome Location
- Chr. 2: 53970838 - 54305300 on Build GRCh38
- UniGene ID
- Hs.516173
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320586.1 |
NP_001307515.1
|
6, 7 |
651 |
|
NM_001320587.1 |
NP_001307516.1
|
5, 6 |
558 |
|
NM_001320588.1 |
NP_001307517.1
|
3, 4 |
352 |
|
NM_001320589.1 |
NP_001307518.1
|
3, 4 |
603 |
|
NM_138448.3 |
NP_612457.1
|
3, 4 |
603 |
|
XM_017005411.1 |
XP_016860900.1
|
7, 8 |
997 |
|
XM_017005412.1 |
XP_016860901.1
|
7, 8 |
997 |
|
GenBank mRNA |
BC012290.1 |
- |
3, 4 |
372 |
|
BF671963.1 |
- |
3, 4 |
351 |
|
BG741807.1 |
- |
5, 6 |
552 |
|
BU543943.1 |
- |
6, 7 |
651 |
|
BU857820.1 |
- |
3, 4 |
461 |
|
X84195.1 |
- |
3, 4 |
352 |
|
View Full Product Details