Gene Transcripts

Gene Symbol
WTAP
Entrez Gene ID
9589
Gene Name
Wilms tumor 1 associated protein
Gene Aliases
Mum2
Chromosome Location
Chr. 6: 159726680 - 159756319 on Build GRCh38
UniGene ID
Hs.446091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270531.1 NP_001257460.1 4 323
NM_001270532.1 NP_001257461.1 4 455
NM_001270533.1 NP_001257462.1 4 323
NM_004906.4 NP_004897.2 4 455
NM_152857.2 NP_690596.1 4 323
NM_152858.2 NP_690597.1 4 455
XM_017011514.1 XP_016867003.1 4 293
XM_017011515.1 XP_016867004.1 2 555
XM_017011516.1 XP_016867005.1 4 293
GenBank mRNA AF374416.1 - 4 336
AJ276706.1 - 3 103
AK127822.1 - 4 334
AK290854.1 - 4 336
BC000383.2 - 4 314
BC004432.1 - 4 314
BC069192.1 - 4 291 4653831
BE906617.1 - 4 440
BG702882.1 - 4 327
BQ948913.1 - 4 261
BX387163.2 - 4 305
CN284477.1 - 4 285
D14661.1 - 4 235
DA138013.1 - 4 455
DA561768.1 - 4 329

View Full Product Details