Gene Transcripts

Gene Symbol
SNX16
Entrez Gene ID
64089
Gene Name
sorting nexin 16
Gene Aliases
Chromosome Location
Chr. 8: 81799583 - 81842326 on Build GRCh38
UniGene ID
Hs.492121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022133.3 NP_071416.2 3 592
NM_152836.2 NP_690049.1 2 346
NM_152837.2 NP_690050.1 2 346
XM_005251282.4 XP_005251339.1 2 386
XM_005251283.2 XP_005251340.1 3 523
GenBank mRNA AF305779.1 - 2 197
AK290903.1 - 2 262
AL833763.1 - 2 253
BC033630.1 - 2 254 5224216
BG700892.1 - 3 613
BG739930.1 - 2 260
CB962009.1 - 2 324

View Full Product Details