Gene Transcripts

Gene Symbol
PATJ
Entrez Gene ID
10207
Gene Name
PATJ, crumbs cell polarity complex component
Gene Aliases
Cipp, INADL, InaD-like, hINADL
Chromosome Location
Chr. 1: 61742477 - 62163920 on Build GRCh38
UniGene ID
Hs.478125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176877.2 NP_795352.2 3 215
XM_005270341.3 XP_005270398.1 3 213
XM_005270347.1 XP_005270404.1 3 213
XM_006710278.3 XP_006710341.1 3 213
XM_011540462.2 XP_011538764.1 3 213
XM_011540463.2 XP_011538765.1 3 401
XM_011540464.2 XP_011538766.1 3 213
XM_011540465.2 XP_011538767.1 3 213
XM_011540466.2 XP_011538768.1 3 213
XM_011540467.2 XP_011538769.1 3 213
XM_011540468.2 XP_011538770.1 3 213
XM_011540469.2 XP_011538771.1 3 213
XM_011540470.1 XP_011538772.1 3 213
XM_016999998.1 XP_016855487.1 3 213
XM_016999999.1 XP_016855488.1 3 213
GenBank mRNA AF397170.1 - 2 101
AJ001306.1 - 3 215
AJ224747.1 - 3 215
AJ224748.1 - 3 215
AK307713.1 - 3 212
AK308428.1 - 3 212
BC142661.1 - 3 198 40148087
BC142723.1 - 3 198 40148887
BC144135.1 - 2 147

View Full Product Details