Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001412.3 | NP_001403.1 | 2 | 287 | |
GenBank mRNA | AK312320.1 | - | 1 | 274 | |
BC000793.1 | - | 2 | 248 | 3451631 | |
BC067851.1 | - | 2 | 274 | 5288453 | |
BC110421.1 | - | 2 | 252 | ||
BT007064.1 | - | 1 | 79 | ||
L18960.1 | - | 2 | 286 |