Gene Transcripts
- Gene Symbol
- SLC29A2
- Entrez Gene ID
- 3177
- Gene Name
- solute carrier family 29 member 2
- Gene Aliases
- DER12, ENT2, HNP36
- Chromosome Location
- Chr. 11: 66362521 - 66372476 on Build GRCh38
- UniGene ID
- Hs.569017
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300868.1 |
NP_001287797.1
|
8, 9 |
1089 |
|
NM_001300869.1 |
NP_001287798.1
|
8, 9 |
1054 |
|
NM_001532.2 |
NP_001523.2
|
6, 7 |
888 |
|
NR_125343.1 |
|
6, 7 |
888 |
|
XM_011544974.1 |
XP_011543276.1
|
8, 9 |
1039 |
|
XM_017017630.1 |
XP_016873119.1
|
6, 7 |
1242 |
|
XM_017017631.1 |
XP_016873120.1
|
6, 7 |
1242 |
|
XM_017017632.1 |
XP_016873121.1
|
8, 9 |
1039 |
|
XM_017017633.1 |
XP_016873122.1
|
6, 7 |
1242 |
|
XM_017017634.1 |
XP_016873123.1
|
6, 7 |
1242 |
|
XM_017017635.1 |
XP_016873124.1
|
6, 7 |
1242 |
|
XM_017017636.1 |
XP_016873125.1
|
6, 7 |
1242 |
|
XM_017017637.1 |
XP_016873126.1
|
6, 7 |
1242 |
|
XM_017017638.1 |
XP_016873127.1
|
4, 5 |
412 |
|
GenBank mRNA |
AF029358.1 |
- |
6, 7 |
662 |
|
AF034102.1 |
- |
6, 7 |
896 |
|
AF401235.1 |
- |
6, 7 |
896 |
|
AK057041.1 |
- |
7, 8 |
879 |
|
BC011387.1 |
- |
7, 8 |
769 |
3872824 |
BC093634.1 |
- |
6, 7 |
708 |
7939479 |
BC143335.1 |
- |
6, 7 |
708 |
|
X86681.1 |
- |
6, 7 |
654 |
|
View Full Product Details