Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001018070.2 | NP_001018080.1 | 3 | 213 | |
NM_020441.2 | NP_065174.1 | 2 | 191 | ||
GenBank mRNA | AK000860.1 | - | 2 | 174 | |
AK295782.1 | - | 2 | 151 | ||
AK309267.1 | - | 2 | 166 | ||
AK315399.1 | - | 2 | 146 | ||
BC006449.1 | - | 2 | 141 | 3958930 | |
BI767141.1 | - | 3 | 189 | ||
DA061767.1 | - | 3 | 213 |