Gene Transcripts

Gene Symbol
FBXL18
Entrez Gene ID
80028
Gene Name
F-box and leucine rich repeat protein 18
Gene Aliases
Fbl18
Chromosome Location
Chr. 7: 5454448 - 5513813 on Build GRCh38
UniGene ID
Hs.623974
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321213.1 NP_001308142.1 2 234
NM_024963.5 NP_079239.3 2 234
XM_011515531.2 XP_011513833.1 2 249
XM_011515532.2 XP_011513834.1 2 249
GenBank mRNA AK057042.1 - 2 188
AK294949.1 - 2 228
BC006426.1 - 2 204 4026501
BC024256.1 - 2 204 3954975
DC307306.1 - 2 234

View Full Product Details