Gene Transcripts

Gene Symbol
CDC26
Entrez Gene ID
246184
Gene Name
cell division cycle 26
Gene Aliases
ANAPC12, APC12, C9orf17
Chromosome Location
Chr. 9: 113267008 - 113275589 on Build GRCh38
UniGene ID
Hs.727648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139286.3 NP_644815.1 3, 4 448
XM_017014573.1 XP_016870062.1 3, 4 415
XM_017014574.1 XP_016870063.1 3, 4 880
XM_017014575.1 XP_016870064.1 3, 4 410
GenBank mRNA AF503918.1 - 1 89
BC042534.1 - 3, 4 403 4751173
BC066300.1 - 3, 4 433 5258904
BF244254.1 - 3, 4 445
CD685602.1 - 3, 4 453

View Full Product Details