Gene Transcripts

Gene Symbol
Kcnb1
Entrez Gene ID
25736
Gene Name
potassium channel, voltage gated Shab-related subfamily B, member 1
Gene Aliases
DRK1PC, Kcr1-1, Kv2.1, Shab
Chromosome Location
Chr. 3: 163850785 - 163935610 on Build Rnor_6.0
UniGene ID
Rn.26724
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013186.1 NP_037318.1 1 247
GenBank mRNA X16476.1 - 1 247

View Full Product Details