Gene Transcripts
- Gene Symbol
- Adra2c
- Entrez Gene ID
- 24175
- Gene Name
- adrenoceptor alpha 2C
- Gene Aliases
- Chromosome Location
- Chr. 14: 80730307 - 80732010 on Build Rnor_6.0
- UniGene ID
- Rn.10297
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138506.1 |
NP_612515.1
|
1 |
1332 |
|
GenBank mRNA |
M58316.1 |
- |
1 |
1332 |
|
View Full Product Details