Gene Transcripts

Gene Symbol
Nampt
Entrez Gene ID
297508
Gene Name
nicotinamide phosphoribosyltransferase
Gene Aliases
Pbef, Pbef1
Chromosome Location
Chr. 6: 52122085 - 52156473 on Build Rnor_6.0
UniGene ID
Rn.203508
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177928.3 NP_808789.1 2 161
GenBank mRNA AB081730.1 - 2 136
AY831728.1 - 2 134
BC085681.1 - 2 161 7192109

View Full Product Details