Gene Transcripts

Gene Symbol
Cryaa
Entrez Gene ID
24273
Gene Name
crystallin, alpha A
Gene Aliases
Acry-1, Crya1
Chromosome Location
Chr. 20: 10438037 - 10442189 on Build Rnor_6.0
UniGene ID
Rn.127769
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289737.1 NP_001276666.1 3, 4 403
NM_012534.4 NP_036666.2 2, 3 379
XM_008772796.1 XP_008771018.1 3, 4 873
GenBank mRNA BC126082.1 - 2, 3 379
M96949.1 - 1, 2 53
M96950.1 - 1, 2 53
U47922.1 - 2, 3 332
V01219.1 - 2, 3 166

View Full Product Details