Gene Transcripts

Gene Symbol
MTMR1
Entrez Gene ID
8776
Gene Name
myotubularin related protein 1
Gene Aliases
Chromosome Location
Chr. X: 150692997 - 150765103 on Build GRCh38
UniGene ID
Hs.347187
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306144.1 NP_001293073.1 2 329
NM_001306145.1 NP_001293074.1 2 329
NM_003828.3 NP_003819.1 2 329
XM_005274765.3 XP_005274822.1 2 291
XM_005274766.3 XP_005274823.1 2 290
XM_005274767.3 XP_005274824.1 2 309
XM_006724855.3 XP_006724918.1 2 329
XM_011531209.2 XP_011529511.1 2 89
XM_011531210.2 XP_011529512.1 2 293
XM_017029920.1 XP_016885409.1 2 291
XM_017029922.1 XP_016885411.1 2 85
XM_017029924.1 XP_016885413.1 2 309
GenBank mRNA AF057354.1 - 2 153
AJ224979.1 - 2 155
AK226109.1 - 2 283
AK295738.1 - 2 124
BC032947.1 - 2 326 4837744
DR000536.1 - 2 331

View Full Product Details