Gene Transcripts

Gene Symbol
Aqp5
Entrez Gene ID
25241
Gene Name
aquaporin 5
Gene Aliases
Chromosome Location
Chr. 7: 141249044 - 141252576 on Build Rnor_6.0
UniGene ID
Rn.10066
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012779.1 NP_036911.1 4 855
GenBank mRNA BC078904.1 - 4 849
BC092572.1 - 4 844 7122974
BC110045.1 - 4 860 7100092
U16245.1 - 4 855

View Full Product Details