Gene Transcripts
- Gene Symbol
- Tnr
- Entrez Gene ID
- 25567
- Gene Name
- tenascin R
- Gene Aliases
- J1NRM
- Chromosome Location
- Chr. 13: 77602249 - 77678385 on Build Rnor_6.0
- UniGene ID
- Rn.11028
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013045.1 |
NP_037177.1
|
1 |
343 |
|
GenBank mRNA |
Z18630.1 |
- |
1 |
343 |
|
View Full Product Details