Gene Transcripts

Gene Symbol
Ppp3ca
Entrez Gene ID
24674
Gene Name
protein phosphatase 3, catalytic subunit, alpha isozyme
Gene Aliases
Calna1
Chromosome Location
Chr. 2: 241909336 - 242186861 on Build Rnor_6.0
UniGene ID
Rn.228648
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017041.1 NP_058737.1 2 371
XM_006233363.2 XP_006233425.1 2 866
XM_006233364.2 XP_006233426.1 2 131
GenBank mRNA AY724517.1 - 2 500
D90035.1 - 2 371
FQ212355.1 - 2 715
M29275.1 - 2 371
X57115.1 - 2 434

View Full Product Details