Gene Transcripts

Gene Symbol
Fdx1
Entrez Gene ID
29189
Gene Name
ferredoxin 1
Gene Aliases
Chromosome Location
Chr. 8: 56373729 - 56393199 on Build Rnor_6.0
UniGene ID
Rn.6946
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017126.1 NP_058822.1 3 444
GenBank mRNA D50436.1 - 3 444
DQ255900.1 - 1 48

View Full Product Details