Gene Transcripts
- Gene Symbol
- Foxa3
- Entrez Gene ID
- 25100
- Gene Name
- forkhead box A3
- Gene Aliases
- Hnf3g
- Chromosome Location
- Chr. 1: 79921292 - 79930219 on Build Rnor_6.0
- UniGene ID
- Rn.10949
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017077.2 |
NP_058773.1
|
2 |
744 |
|
GenBank mRNA |
L09648.1 |
- |
2 |
745 |
|
View Full Product Details