Gene Transcripts
- Gene Symbol
- Areg
- Entrez Gene ID
- 29183
- Gene Name
- amphiregulin
- Gene Aliases
- Chromosome Location
- Chr. 14: 18521921 - 18531179 on Build Rnor_6.0
- UniGene ID
- Rn.10568
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017123.1 |
NP_058819.1
|
3 |
502 |
|
GenBank mRNA |
X55183.1 |
- |
3 |
502 |
|
View Full Product Details