Gene Transcripts

Gene Symbol
Zfr2
Entrez Gene ID
103406
Gene Name
zinc finger RNA binding protein 2
Gene Aliases
2010013I23Rik, 9130206N08Rik, AW045600, mKIAA1086
Chromosome Location
Chr. 10: 81233163 - 81252123 on Build GRCm38
UniGene ID
Mm.158783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034895.3 NP_001030067.2 18 3007
GenBank mRNA AK033649.1 - 11 1756
AK122438.1 - 8 5642
BC072571.1 - 16 4193

View Full Product Details