Gene Transcripts

Gene Symbol
Wdr90
Entrez Gene ID
106618
Gene Name
WD repeat domain 90
Gene Aliases
3230401M21Rik, AI551153, mKIAA1924
Chromosome Location
Chr. 17: 25844734 - 25861515 on Build GRCm38
UniGene ID
Mm.35470
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163766.1 NP_001157238.1 22 3149
GenBank mRNA AK014328.1 - 18 2250
AK129465.1 - 21 3064
AK154827.1 - 20 3316

View Full Product Details