Gene Transcripts

Gene Symbol
Cd69
Entrez Gene ID
12515
Gene Name
CD69 antigen
Gene Aliases
5830438K24Rik, AI452015, AIM, VEA
Chromosome Location
Chr. 6: 129267324 - 129275369 on Build GRCm38
UniGene ID
Mm.74745
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033122.3 NP_001028294.1 5 1177
GenBank mRNA AK017979.1 - 5 1176
AK151076.1 - 5 1178
AK153224.1 - 5 1178

View Full Product Details