Gene Transcripts

Gene Symbol
Fbrs
Entrez Gene ID
14123
Gene Name
fibrosin
Gene Aliases
Fbs, Fbs1
Chromosome Location
Chr. 7: 127477246 - 127491513 on Build GRCm38
UniGene ID
Mm.323505
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010183.1 NP_034313.1 10 781
GenBank mRNA AK037952.1 - 6 560
BC027193.1 - 8 652
CF532387.1 - 4 451

View Full Product Details