Gene Transcripts

Gene Symbol
Fbrs
Entrez Gene ID
14123
Gene Name
fibrosin
Gene Aliases
Fbs, Fbs1
Chromosome Location
Chr. 7: 127477246 - 127491513 on Build GRCm38
UniGene ID
Mm.323505
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010183.1 NP_034313.1 11 1733
GenBank mRNA AK037952.1 - 7 1512
BC027193.1 - 9 1604

View Full Product Details