Gene Transcripts
- Gene Symbol
- Cma2
- Entrez Gene ID
- 545055
- Gene Name
- chymase 2, mast cell
- Gene Aliases
- Mcp10
- Chromosome Location
- Chr. 14: 55950806 - 55974031 on Build GRCm38
- UniGene ID
- Mm.378966
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M68899.1 |
- |
4 |
818 |
|
View Full Product Details