Gene Transcripts

Gene Symbol
Gm9
Entrez Gene ID
194854
Gene Name
predicted gene 9
Gene Aliases
Chromosome Location
Chr. X: 37208502 - 37211041 on Build GRCm38
UniGene ID
Mm.296429
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033234.2 NP_001028406.1 4 799
GenBank mRNA AK145837.1 - 4 800
BC048624.1 - 4 763

View Full Product Details