Gene Transcripts

Gene Symbol
Kntc1
Entrez Gene ID
208628
Gene Name
kinetochore associated 1
Gene Aliases
D330012D13
Chromosome Location
Chr. 5: 123749689 - 123821593 on Build GRCm38
UniGene ID
Mm.441089
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042421.1 NP_001035886.1 3 326
GenBank mRNA AK172905.1 - 3 300
BY010626.1 - 3 327

View Full Product Details