Gene Transcripts

Gene Symbol
Chadl
Entrez Gene ID
214685
Gene Name
chondroadherin-like
Gene Aliases
AY100452, D930017K21Rik
Chromosome Location
Chr. 15: 81686166 - 81697376 on Build GRCm38
UniGene ID
Mm.393287
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164320.1 NP_001157792.1 6 2393
GenBank mRNA AK139481.1 - 3 957

View Full Product Details