Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033269.3 | NP_001028441.1 | 7 | 1801 | |
NM_001039683.2 | NP_001034772.1 | 7 | 1763 | ||
NM_001286178.1 | NP_001273107.1 | 7 | 1730 | ||
NM_001286179.1 | NP_001273108.1 | 7 | 1733 | ||
NM_001286180.1 | NP_001273109.1 | 7 | 1692 | ||
GenBank mRNA | AK139577.1 | - | 7 | 1395 | |
DQ279473.1 | - | 7 | 1796 |