Gene Transcripts

Gene Symbol
Slc44a5
Entrez Gene ID
242259
Gene Name
solute carrier family 44, member 5
Gene Aliases
AA117069, Gm422
Chromosome Location
Chr. 3: 153973436 - 154271747 on Build GRCm38
UniGene ID
Mm.239962
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081263.1 NP_001074732.1 16 1895
GenBank mRNA BC068154.1 - 3 680
BC086641.1 - 16 1895 6417957

View Full Product Details