Gene Transcripts

Gene Symbol
Sprtn
Entrez Gene ID
244666
Gene Name
SprT-like N-terminal domain
Gene Aliases
Gm505
Chromosome Location
Chr. 8: 124897771 - 124906164 on Build GRCm38
UniGene ID
Mm.482915
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111141.1 NP_001104611.1 5 1482
GenBank mRNA AK154380.1 - 5 1485
BC158088.1 - 5 1337 9007378

View Full Product Details