Gene Transcripts

Gene Symbol
Dpcr1
Entrez Gene ID
268949
Gene Name
diffuse panbronchiolitis critical region 1 (human)
Gene Aliases
Gm630
Chromosome Location
Chr. 17: 35635755 - 35643695 on Build GRCm38
UniGene ID
Mm.296301
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033366.3 NP_001028538.1 2 831
GenBank mRNA AK131887.1 - 2 833
AK164929.1 - 2 832
BC150798.1 - 2 824 9087709

View Full Product Details