Gene Transcripts

Gene Symbol
Ksr2
Entrez Gene ID
333050
Gene Name
kinase suppressor of ras 2
Gene Aliases
Chromosome Location
Chr. 5: 117413770 - 117775004 on Build GRCm38
UniGene ID
Mm.437947
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114545.2 NP_001108017.2 7, 8 2149
GenBank mRNA BC068233.1 - 5, 6 934
DQ531035.1 - 7, 8 1310
KJ719253.1 - 3, 4 265

View Full Product Details