Gene Transcripts

Gene Symbol
Gipr
Entrez Gene ID
381853
Gene Name
gastric inhibitory polypeptide receptor
Gene Aliases
GIP-R, Gm1081, Gm160
Chromosome Location
Chr. 7: 19156063 - 19170248 on Build GRCm38
UniGene ID
Mm.333633
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080815.1 NP_001074284.1 10 1064
GenBank mRNA AK133386.1 - 5 792
AK146552.1 - 9 1270

View Full Product Details