Gene Transcripts
- Gene Symbol
- Trim34b
- Entrez Gene ID
- 434218
- Gene Name
- tripartite motif-containing 34B
- Gene Aliases
- Gm15134, OTTMUSG00000019188, Trim34-2
- Chromosome Location
- Chr. 7: 104329471 - 104336617 on Build GRCm38
- UniGene ID
- Mm.485757
- Species
- Mus musculus
- Gene Symbol
- Trim34a
- Entrez Gene ID
- 94094
- Gene Name
- tripartite motif-containing 34A
- Gene Aliases
- Trim34, Trim34-1
- Chromosome Location
- Chr. 7: 104244437 - 104262239 on Build GRCm38
- UniGene ID
- Mm.326945
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF220140.1 |
- |
|
1620 |
|
View Full Product Details