Gene Transcripts

Gene Symbol
Hmcn1
Entrez Gene ID
545370
Gene Name
hemicentin 1
Gene Aliases
EG545370, Gm201
Chromosome Location
Chr. 1: 150562500 - 150993687 on Build GRCm38
UniGene ID
Mm.379425
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024720.3 NP_001019891.2 28 4715

View Full Product Details