Gene Transcripts

Gene Symbol
Gpr52
Entrez Gene ID
620246
Gene Name
G protein-coupled receptor 52
Gene Aliases
EG620246
Chromosome Location
Chr. 1: 160576668 - 160577753 on Build GRCm38
UniGene ID
Mm.478780
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146330.1 NP_001139802.1 1 250

View Full Product Details